Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLP Antibody (55N1E3), Novus Biologicals™
SDP

Catalog No. NBP227323 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227323 0.1 mg
NBP227323SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227323 Supplier Novus Biologicals Supplier No. NBP227323
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSLP Monoclonal specifically detects TSLP in Human samples. It is validated for Western Blot.

Specifications

Antigen TSLP
Applications Western Blot
Classification Monoclonal
Clone 55N1E3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2ug/ml
Gene Accession No. Q969D9
Gene Alias thymic stromal lymphopoietin
Gene Symbols TSLP
Host Species Mouse
Immunogen amino acids (27-158) MDYDFTNCDFEkIkAAYLSTISkDLITYMSGTkSTEFNNTVSCSNRPHCLTEIQSLTFNP∼TAGCASLAkEMFAMkTkAALAIWCPGYSETQINATQAMkkRRkRkVTTNkCLEQVSQLQG∼LWRRFNRPLLkQQL
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85480
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.