Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSLP Monoclonal Antibody (152640)

Rat Monoclonal Antibody
$603.00 - $938.00
Specifications
| Antigen | TSLP |
|---|---|
| Clone | 152640 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA523779
|
Invitrogen™
MA523779 |
500 μg |
Each for $938.00
|
|
|||||
PIMA523875
![]() |
Invitrogen™
MA523875 |
100 μg |
Each for $603.00
|
|
|||||
Description
In Western blots, this antibody does not cross-react with recombinant mouse IL-7 or recombinant human TSLP. Reconstitute at 0.5 mg/mL in sterile PBS.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.Spécifications
| TSLP | |
| 0.5 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 5% trehalose and No Preservative | |
| Q9JIE6 | |
| 53603 | |
| Sf 21-derived recombinant mouse TSLP Tyr20-Glu140. | |
| Antibody |
| 152640 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Lyophilized | |
| IgG2a | |
| Mouse | |
| TSLP | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
| TSLP | |
| Primary | |
| Protein A/G |
missing translation for 'spotOppurtunityImprovement'missing translation for 'shareContentCorrection'
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'