Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSLP Receptor, Mouse anti-Human, Clone: 1A6, Millipore Sigma™
Mouse Monoclonal Antibody
$482.80
Specifications
Antigen | TSLP Receptor |
---|---|
Clone | 1A6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2042100
|
MilliporeSigma
MABF2042100UG |
100 μg |
Each of 1 for $482.80
|
|
|||||
Description
Anti-TSLP Receptor, clone 1A6, Cat. No. MABF2042, is a mouse monoclonal antibody that detects TSLP receptor and has been tested for use in Flow Cytometry.
Cytokine receptor-like factor 2 (UniProt: Q9HC73; also known as Cytokine receptor-like 2, IL-XR, Thymic stromal lymphopoietin protein receptor, TSLP receptor) is encoded by the CRLF2 (also known as CRL2, ILXR, TSLPR) gene (Gene ID: 64109) in human. TSLP receptor is a single-pass type I membrane protein that serves as a receptor for thymic stromal lymphopoietin. It forms a functional complex with TSLP and IL7R to stimulate cell proliferation via activation of STAT3 and STAT5. It is primarily expressed in dendrites and monocytes and its expression has also been reported in heart, skeletal muscle, kidney and liver. Its levels are up-regulated in activated peripheral monocytes and THP-1 cells. TSLP receptor is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 23-231), a transmembrane domain (aa 232-252), and a cytoplasmic domain (aa 253-371). It also has a WSXWS motif (aa 200-204) that is essential for proper protein folding and efficient intracellular transport and cell-surface binding. In addition, its Box 1 motif (aa 261-269) is reported to be important for its association with JAKs. Three isoforms of TSLP receptors have been described that are produced by alternative splicing.Specifications
TSLP Receptor | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_071431.2 | |
Fc-fusion protein of ectodomain region of human TSLP receptor protein. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
1A6 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cytokine receptor-like factor 2;Cytokine receptor-like 2;IL-XR;Thymic stromal lymphopoietin protein receptor;TSLPR | |
CRLF2;CRL2;ILXR;TSLPR | |
IgG2a λ | |
Protein G purified | |
Clone 1A6 specifically detects human TSLP receptor protein. It targets an epitope with in the extracellular domain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title