Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSLP Receptor, Rat anti-Mouse, Clone: 22H9, Millipore Sigma™
Rat Monoclonal Antibody
$192.34 - $482.80
Specifications
Antigen | TSLP Receptor |
---|---|
Clone | 22H9 |
Dilution | Flow Cytometry Analysis: A representative lot detected TSLP receptor in Flow Cytometry applications (Bell, B.D., et. al. (2013). Nat Immunol. 14(4):364-71; Isaksen, D.E., et. al. (2002). J Immunol. 168(7):3288-94; Iseki, M., et. al. (2012). Int Immunol. 24(3):183-95; Kashiwagi, M., et. al. (2017). Nat Immunol. 18(3):334-343). |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF196625
|
MilliporeSigma
MABF196625UG |
25 μg |
Each for $192.34
|
|
|||||
MABF1966100
|
MilliporeSigma
MABF1966100UG |
100 μg |
Each for $482.80
|
|
|||||
Description
Anti-TSLP Receptor, clone 22H9, Cat. No. MABF1966, is a rat monoclonal antibody that detects TSLP receptor and has been tested for use in Flow Cytometry.
Cytokine receptor-like factor 2 (UniProt: Q8CII9; also known as Cytokine receptor-like molecule 2, CRLM-2, Thymic stromal lymphopoietin protein receptor, TSLP receptor, Type I cytokine receptor delta 1) is encoded by the Crlf2 (also known as rlm2, Tpte2, Tslpr) gene (Gene ID: 57914) in murine species. TSLP receptor is a single-pass type I membrane protein that serves as a receptor for thymic stromal lymphopoietin. It is expressed in liver, lung, testis, heart, brain, spleen, thymus, and bone marrow. Higher expression levels are observed in progenitors and myeloid cells. Its levels can be up-regulated in the myeloid 32D cell line by granulocyte colony-stimulating factor. TSLP receptor forms a functional complex with TSLP and IL7R, which is capable of stimulating cell proliferation through activation of STAT3 and STAT5. It has also been implicated in the development of the hematopoietic system. It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form. The mature form contains an extracellular domain (aa 20-232), a transmembrane domain (aa 233-253), and a cytoplasmic domain (aa 254-359). It contains WSXWS motif (aa 201-205), which is essential for proper protein folding and efficient intracellular transport and cell-surface binding. It also has a Box 1 motif (aa 262-270) that is required for JAK interaction and activation.Specifications
TSLP Receptor | |
Flow Cytometry Analysis: A representative lot detected TSLP receptor in Flow Cytometry applications (Bell, B.D., et. al. (2013). Nat Immunol. 14(4):364-71; Isaksen, D.E., et. al. (2002). J Immunol. 168(7):3288-94; Iseki, M., et. al. (2012). Int Immunol. 24(3):183-95; Kashiwagi, M., et. al. (2017). Nat Immunol. 18(3):334-343). | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
Cytokine receptor-like factor 2;Cytokine receptor-like molecule 2;CRLM-2;Thymic stromal lymphopoietin protein receptor;Type I cytokine receptor delta 1 | |
Crlf2;rlm2;Tpte2;Tslpr | |
IgG2a κ | |
Protein G purified | |
Clone 22H9 is a rat monoclonal antibody that detects Thymic stromal lymphopoietin protein receptor (TSLPR) in murine species. It targets an epitope with in the extracellular domain. |
22H9 | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Inflammation & Immunology | |
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001158207.1 | |
His-tagged murine recombinant TSLP Receptor Protein. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title