Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLPR/CRLF2 Antibody (147036), Biotin, Novus Biologicals™
SDP

Catalog No. NB295345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295345 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295345 Supplier Novus Biologicals Supplier No. FAB981B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSLPR/CRLF2 Monoclonal antibody specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen TSLPR/CRLF2
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 147036
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TSLPR, Gly25-Lys231, Accession # Q9HC73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64109
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.