Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLPR/CRLF2 Antibody (59N5G4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP227331UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227331UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227331UV Supplier Novus Biologicals Supplier No. NBP227331UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSLPR/CRLF2 Monoclonal specifically detects TSLPR/CRLF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TSLPR/CRLF2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 59N5G4
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. Q9HC73
Gene Alias CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR
Gene Symbols CRLF2
Host Species Mouse
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64109
Test Specificity TSLPR is a glycoprotein with two major isoforms of 359 amino acids and 255 amino acids. In western blotting, bands may be visualized between 27 kD and ∼50 kD.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.