Learn More
Description
Specifications
Specifications
| Antigen | TSLPR/CRLF2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 59N5G4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin |
| Gene Accession No. | Q9HC73 |
| Gene Alias | CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR |
| Gene Symbols | CRLF2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
