Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN1 Antibody (819202), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB310540 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310540 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310540 Supplier Novus Biologicals Supplier No. FAB9186PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TSPAN1
Applications Flow Cytometry
Classification Monoclonal
Clone 819202
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TSPAN1, Met1-Gln241, Accession # O60635
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10103
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.