Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN1 Rabbit anti-Human, Clone: 10, Novus Biologicals™
SDP

Catalog No. NB021798 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021798 50 μg
NB021797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021798 Supplier Novus Biologicals Supplier No. NBP29016750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TSPAN1 Monoclonal specifically detects TSPAN1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TSPAN1
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:5000-1:20000
Formulation 0.2 um filtered solution in PBS
Gene Alias NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1
Host Species Rabbit
Immunogen Recombinant Human TSPAN1 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10103
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.