Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN1 Rabbit anti-Human, Clone: 2022-04-12 00:00:00, Novus Biologicals™
SDP

Catalog No. NB154195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154195 0.2 mg
NB154194 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB154195 Supplier Novus Biologicals Supplier No. NBP3120610.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen TSPAN1
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 2022-04-12 00:00:00
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1
Host Species Rabbit
Immunogen This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10103
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.