Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN1 Rabbit anti-Human, Clone: 2022-04-12 00:00:00, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | TSPAN1 |
---|---|
Clone | 2022-04-12 00:00:00 |
Concentration | 1 mg/ml |
Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | ELISA, Immunohistochemistry, Immunofluorescence |
Description
TSPAN1 Monoclonal antibody specifically detects TSPAN1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
TSPAN1 | |
1 mg/ml | |
ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
NET1, NET-1, tetraspan 1, Tetraspan NET-1, tetraspanin 1, Tetraspanin TM4-C, tetraspanin-1, TM4C, TM4SF, TSPAN-1 | |
This antibody was raised by immunising BALB/c mice with the major extracellular domain of tetraspanin 1. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
2022-04-12 00:00:00 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
10103 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title