Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN12 Antibody (921938), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345936 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345936 Supplier Novus Biologicals Supplier No. FAB8910MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN12 Monoclonal specifically detects TSPAN12 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen TSPAN12
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 921938
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, NET-2, ten-eleven translocation-1, tetraspanin-12
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TSPAN12, Met1-Leu305
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23554
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.