Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN2 Antibody (822509), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311088 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311088 Supplier Novus Biologicals Supplier No. FAB7876APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN2 Monoclonal specifically detects TSPAN2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TSPAN2
Applications Flow Cytometry
Classification Monoclonal
Clone 822509
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias FLJ12082, NET3, new EST tetraspan 3, tetraspan 2, Tetraspan NET-3, tetraspan TM4SF, tetraspanin 2, tetraspanin 2 isoform, tetraspanin-2, TSN2, TSPAN-2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TSPAN2, Met1-Ile221, Accession # O60636
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10100
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.