Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095907 100 μL
NB095908 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095907 Supplier Novus Biologicals Supplier No. NBP304386100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPAN3 Polyclonal specifically detects TSPAN3 in Human samples. It is validated for Western Blot.

Specifications

Antigen TSPAN3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias tetraspan TM4SF, tetraspanin 3, Tetraspanin TM4-A, tetraspanin-3, TM4-A, TM4SF8, Transmembrane 4 superfamily member 8tetraspan 3, TSPAN-3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 105-210 of human TSPAN3 (NP_005715.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10099
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.