Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB301761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301761 100 μL
NB159079 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB301761 Supplier Novus Biologicals Supplier No. NBP314797100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPAN3 Polyclonal antibody specifically detects TSPAN3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TSPAN3
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:100-1:500
Formulation PBS, pH7.0
Gene Alias tetraspan TM4SF, tetraspanin 3, Tetraspanin TM4-A, tetraspanin-3, TM4-A, TM4SF8, Transmembrane 4 superfamily member 8tetraspan 3, TSPAN-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TSPAN3 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 10099
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.