Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN32/TSSC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP162646
Description
TSPAN32/TSSC6 Polyclonal specifically detects TSPAN32/TSSC6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TSPAN32/TSSC6 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ART1, FLJ17158, FLJ97586, MGC22455, pan-hematopoietic expression, PHEMXPHMX, Protein Phemx, tetraspanin 32, tetraspanin-32, tspan-32, TSSC6pan-hematopoietic expression protein, tumor-suppressing STF cDNA 6, tumor-suppressing subchromosomal transferable fragment cDNA 6, tumor-suppressing subtransferable candidate 6 | |
Rabbit | |
31 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q96QS1-2 | |
TSPAN32 | |
Synthetic peptides corresponding to TSPAN32(tetraspanin 32) The peptide sequence was selected from the middle region of TSPAN32. Peptide sequence YEQAMKGTSHVRRQELAAIQDVFLCCGKKSPFSRLGSTEADLCQGEEAAR. | |
Protein A purified | |
RUO | |
10077 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction