Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN7/TM4SF2 Antibody (B2D), FITC, Novus Biologicals™
SDP

Catalog No. NB002910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002910 Supplier Novus Biologicals Supplier No. NBP275971F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSPAN7/TM4SF2 Monoclonal specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen TSPAN7/TM4SF2
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone B2D
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7
Gene Symbols TSPAN7
Host Species Mouse
Immunogen Human T-ALL cell line THP-6
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7102
Test Specificity The antibody QBEnd-10 reacts with Class II epitope on CD34 (Mucosialin), a 110-115 kDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells. This antibody has been also used as an endothelial marker.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.