Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN8/TM4SF3 Antibody (458811), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347602 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347602 Supplier Novus Biologicals Supplier No. FAB4734MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TSPAN8/TM4SF3 Monoclonal antibody specifically detects TSPAN8/TM4SF3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen TSPAN8/TM4SF3
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 458811
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CO-029, tetraspanin 8, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029
Host Species Rat
Immunogen NS0 mouse myeloma cell line transfected with human TSPAN8, Met1-Lys237 (Ile35Val), Accession # AAH05246
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7103
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.