Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPY1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343781 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343781 Supplier Novus Biologicals Supplier No. NBP241198MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPY1 Polyclonal antibody specifically detects TSPY1 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen TSPY1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Cancer/testis antigen 78, CT78DYS14, testis specific protein, Y-linked, testis specific protein, Y-linked 1, testis-specific Y-encoded protein 1, TSPYpJA923
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid peptide near the carboxy terminus of human TSPY1L. The immunogen is located within the last 50 amino acids of TSPY1L. Amino Acid Squence: CRNPLQYYKRMKPPEEGTETS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7258
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.