Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TST Mouse anti-Human, Mouse, Rat, Clone: 2A6C5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TST |
---|---|
Clone | 2A6C5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6461
![]() ![]() |
Proteintech Group Inc
660181IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The product of this gene is a mitochondrial matrix enzyme that is encoded by the nucleus. It may play roles in cyanide detoxification, the formation of iron-sulfur proteins, and the modification of sulfur-containing enzymes. The gene product contains two highly conservative domains (rhodanese homology domains), suggesting these domains have a common evolutionary origin.Specifications
TST | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RDS, Rhodanese, Thiosulfate sulfurtransferase, TST | |
TST | |
IgG2a | |
Protein A | |
Antibody |
2A6C5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P24329, P52196, Q16762 | |
22117, 25274, 7263 | |
TST Fusion Protein Ag9235 | |
Primary | |
-20°C | |
TST |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title