Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTBK1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB136453 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136453 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136453 Supplier Novus Biologicals Supplier No. NBP306356AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTBK1 Polyclonal specifically detects TTBK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TTBK1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BDTK, Brain-derived tau kinase, EC 2.7.11, EC 2.7.11.1, KIAA1855tau-tubulin kinase 1, RP3-330M21.4, tau tubulin kinase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TTBK1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 84630
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.