Learn More
Description
Specifications
Specifications
| Antigen | TTC11 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA reported in scientific literature (PMID 27494529), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34999205), Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Wang et al (2009)), KnockDown Validated |
| Formulation | Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q9Y3D6 |
| Gene Alias | CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 |
| Gene Symbols | FIS1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
