Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC11 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056646T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056646T 0.05 mL
NB10056646 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056646T Supplier Novus Biologicals Supplier No. NB10056646SS

Rabbit Polyclonal Antibody

TTC11 Polyclonal specifically detects TTC11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen TTC11
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA reported in scientific literature (PMID 27494529), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34999205), Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Wang et al (2009)), KnockDown Validated
Formulation Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y3D6
Gene Alias CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11
Gene Symbols FIS1
Host Species Rabbit
Immunogen Amino acids 11-25 of human Fis1 protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51024
Test Specificity The NCBI accession number for Fis1 is NP_057152. The amino acid sequence used as immunogen is 93% homologous in dog, 73% in mouse and rat, and 66% in chimp (a 512 AA protein).
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.