Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$485.00

Specifications

Antigen TTC11
Dilution Western Blot 1:500 - 1:1000, ELISA reported in scientific literature (PMID 27494529), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34999205), Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Wang et al (2009)), KnockDown Validated
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056646
SDP
View Documents
Novus Biologicals
NB10056646
0.2 mL
Each for $485.00
Only null left
Add to Cart
 
NB10056646T
SDP
View Documents
Novus Biologicals
NB10056646SS
0.05 mL N/A N/A N/A
Description

Description

TTC11 Polyclonal specifically detects TTC11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

TTC11
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
Rabbit
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors
Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11
FIS1
IgG
Unpurified
The NCBI accession number for Fis1 is NP_057152. The amino acid sequence used as immunogen is 93% homologous in dog, 73% in mouse and rat, and 66% in chimp (a 512 AA protein).
Western Blot 1:500 - 1:1000, ELISA reported in scientific literature (PMID 27494529), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34999205), Immunohistochemistry-Paraffin 1:100. Use reported in scientific literature (Wang et al (2009)), KnockDown Validated
Polyclonal
Antisera
RUO
Human, Mouse, Rat
Q9Y3D6
51024
Amino acids 11-25 of human Fis1 protein were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.