Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC14 Antibody (OTI9E11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N274655A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274655A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274655A405 Supplier Novus Biologicals Supplier No. NBP274655AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTC14 Monoclonal specifically detects TTC14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TTC14
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI9E11
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp313M1015, DRDL5813, KIAA1980FLJ00166, PRO19630, tetratricopeptide repeat domain 14, tetratricopeptide repeat protein 14, TPR repeat protein 14
Gene Symbols TTC14
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 429-770 of human TTC14 (NP_597719) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 151613.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.