Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15795820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15795820 20 μL
NBP157958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15795820 Supplier Novus Biologicals Supplier No. NBP15795820UL

Rabbit Polyclonal Antibody

TTC17 Polyclonal specifically detects TTC17 in Human samples. It is validated for Western Blot.

Specifications

Antigen TTC17
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96AE7
Gene Alias DKFZp686D20222, FLJ10890, FLJ13099, FLJ23673, tetratricopeptide repeat domain 17, tetratricopeptide repeat protein 17, TPR repeat protein 17
Gene Symbols TTC17
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TTC17(tetratricopeptide repeat domain 17) The peptide sequence was selected from the N terminal of TTC17. Peptide sequence HNKEDPDCIKAKVPLGDLDLYDGTYITLESKDISPEDYIDTESPVPPDPE.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55761
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.