Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC3 Antibody, Novus Biologicals™
SDP

Catalog No. NB395064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395064 25 μL
NBP255316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB395064 Supplier Novus Biologicals Supplier No. NBP25531625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC3 Polyclonal specifically detects TTC3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TTC3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias DCRR1tetratricopeptide repeat protein 3 (TPR repeat protein D)10RNF105DKFZp686M0150, E3 ubiquitin-protein ligase TTC3, EC 6.3.2.-, Protein DCRR1, RING finger protein 105, tetratricopeptide repeat domain 3, Tetratricopeptide repeat protein 3, TPR repeat protein 3, TPR repeat protein D, TPRDI, TPRDII, TPRDTPRDIII
Gene Symbols TTC3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KPGSEDANYKRVSCNSPKPVLEDVKPTYWAQSHLVTGYCTYLPFQRFDITQTPPAYINVLPGLPQYTSIYTPLASLSPEYQLPRSVPVVPSFVAND
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7267
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.