Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC32 Antibody (OTI1F7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N274656PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274656PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274656PEC7 Supplier Novus Biologicals Supplier No. NBP274656PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTC32 Monoclonal specifically detects TTC32 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TTC32
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1F7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias tetratricopeptide repeat domain 32, tetratricopeptide repeat protein 32, TPR repeat protein 32
Gene Symbols TTC32
Host Species Mouse
Immunogen Full length recombinant protein of human TTC32 (NP_001008238) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 130502.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.