Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC33 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB042547 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042547 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042547 Supplier Novus Biologicals Supplier No. NBP297329AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC33 Polyclonal specifically detects TTC33 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TTC33
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TTC33 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23548
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.