Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC36 Antibody, Novus Biologicals™
SDP

Catalog No. NB287389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287389 Supplier Novus Biologicals Supplier No. NBP323480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC36 Polyclonal antibody specifically detects TTC36 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TTC36
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 0.5-1 ul/mg of lysate
Formulation PBS, pH 7.0
Gene Alias HBP21, tetratricopeptide repeat domain 36, tetratricopeptide repeat protein 36
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TTC36 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143941
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.