Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259139 Supplier Novus Biologicals Supplier No. NBP259139

Rabbit Polyclonal Antibody

TTC4 Polyclonal specifically detects TTC4 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TTC4
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias DKFZp781B0622, FLJ41930, MGC5097, tetratricopeptide repeat domain 4, tetratricopeptide repeat protein 4, TPR repeat protein 4
Gene Symbols TTC4
Host Species Rabbit
Immunogen Maps to a region between 1 and 50
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7268
Target Species Human
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.