Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF1 Mouse anti-Human, Porcine, Clone: 2C8F3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TTF1 |
---|---|
Clone | 2C8F3 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6474
![]() ![]() |
Proteintech Group Inc
660341IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
TTF1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BHC, Homeobox protein Nkx 2.1, NK 2, NK2 homeobox 1, NKX2 1, NKX2.1, NKX2-1, NKX2A, TEBP, Thyroid nuclear factor 1, Thyroid transcription factor 1, TITF1, TTF 1, TTF1 | |
NKX2-1 | |
IgG3 | |
Protein A | |
Antibody |
2C8F3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P43699 | |
100154384, 7080 | |
Peptide | |
Primary | |
-20°C | |
NKX2-1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title