Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB047525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047525 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047525 Supplier Novus Biologicals Supplier No. NBP297769AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTF2 Polyclonal specifically detects TTF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen TTF2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TTF2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.