Learn More
Description
Specifications
Specifications
| Antigen | TTF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.6.1, EC 3.6.4.-, F2, HuF2lodestar protein, human factor 2, Lodestar homolog, RNA polymerase II termination factor, Transcription release factor 2, transcription termination factor 2, transcription termination factor, RNA polymerase II |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TTF2 fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
