Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTI1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP179035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP179035 Supplier Novus Biologicals Supplier No. NBP179035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTI1 Polyclonal specifically detects TTI1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen TTI1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-10ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055472.1
Gene Alias KIAA0406SMG10, Protein SMG10, Tel two interacting protein 1, Tel2 interacting protein 1 homolog (S. pombe), TEL2-interacting protein 1 homolog
Gene Symbols TTI1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1039 and 1089 of human Tel2 Interacting Protein 1 using the numbering given in entry NP_055472.1 (GeneID 9675).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9675
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.