Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTL Antibody (OTI3G5), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB27468A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB27468A488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB27468A488 Supplier Novus Biologicals Supplier No. NBP274668AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTL Monoclonal specifically detects TTL in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TTL
Applications Western Blot
Classification Monoclonal
Clone OTI3G5
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2.25,2410003M22Rik, MGC46235, tubulin tyrosine ligase, tubulin-tyrosine ligase, tubulin--tyrosine ligase
Gene Symbols TTL
Host Species Mouse
Immunogen Full length recombinant protein of human TTL (NP_714923) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 150465.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.