Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTLL5 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290084 Supplier Novus Biologicals Supplier No. NBP232091F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTLL5 Polyclonal antibody specifically detects TTLL5 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TTLL5
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias STAMP, tubulin tyrosine ligase-like family, member 5
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1231 to 1281 of human SRC1 and TIF2-Associated Modulatory Protein using the numbering given in entry NP_055887.3 (GeneID 23093).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23093
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.