Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTLL5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232086 Supplier Novus Biologicals Supplier No. NBP232086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTLL5 Polyclonal specifically detects TTLL5 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen TTLL5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_055887.3
Gene Alias STAMP, tubulin tyrosine ligase-like family, member 5
Gene Symbols TTLL5
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 825 to 875 of human SRC1 and TIF2-Associated Modulatory Protein using the numbering given in entry NP_055887.3 (GeneID 23093).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23093
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.