Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TTNPB
GREENER_CHOICE

Catalog No. p-4785701
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Retinoic acid analog; RAR agonist

Extremely potent analog of retinoic acid, selective for the retinoic acid receptor (RAR) subtype. Enhances reprogramming efficiency in chemically induced pluripotent stem cells (CiPSCs).

Chemical Identifiers

CAS 71441-28-6
Molecular Formula C24H28O2

Specifications

Quantity 10 mg
Formula Weight 348.48
Purity >98%
Molecular Weight (g/mol) 348.486
Chemical Name or Material TTNPB
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.