Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TTP 22
GREENER_CHOICESDP

Catalog No. 443210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-3210 10 mg
44-325-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-3210 Supplier Tocris Bioscience™ Supplier No. 4432/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity, selective CK2 inhibitor

High affinity, ATP-competitive casein kinase 2 (CK2) inhibitor (IC50 = 0.1 μM, Ki = 40 nM). Displays selectivity for CK2 over JNK3, ROCK1 and MET (no inhibitory effects at 10 μM).

Chemical Identifiers

CAS 329907-28-0
Molecular Formula C16H14N2O2S2

Specifications

Quantity 10 mg
Formula Weight 330.42
Purity >98%
Molecular Weight (g/mol) 330.42
Chemical Name or Material TTP 22
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.