Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTYH2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435967 0.025 mg
NBP241177 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435967 Supplier Novus Biologicals Supplier No. NBP2411770.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTYH2 Polyclonal specifically detects TTYH2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen TTYH2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias C17orf29, hTTY2, Protein Tweety Homolog 2, Tweety (Drosophila) Homolog 2, Tweety Family Member 2, Tweety Homolog 2, Tweety Homolog 2 (Drosophila)
Gene Symbols TTYH2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human TTYH2. The immunogen is located within amino acids 60 - 110 of TTYH2. Amino Acid Squence: RRDDAVQTKQHHS
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94015
Test Specificity At least four isoforms of TTYH2 are known to exist; this antibody will only detect the two longest isoforms. This antibody is predicted to not cross-react with other TTYH family members.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.