Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTYH2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | TTYH2 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TTYH2 Polyclonal specifically detects TTYH2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
TTYH2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
94015 | |
Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human TTYH2. The immunogen is located within amino acids 60 - 110 of TTYH2. Amino Acid Squence: RRDDAVQTKQHHS | |
Primary | |
At least four isoforms of TTYH2 are known to exist; this antibody will only detect the two longest isoforms. This antibody is predicted to not cross-react with other TTYH family members. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
C17orf29, hTTY2, Protein Tweety Homolog 2, Tweety (Drosophila) Homolog 2, Tweety Family Member 2, Tweety Homolog 2, Tweety Homolog 2 (Drosophila) | |
TTYH2 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title