Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA6 Antibody (OTI1D8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274692A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274692A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274692A32 Supplier Novus Biologicals Supplier No. NBP274692AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUBA6 Monoclonal specifically detects TUBA6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TUBA6
Applications Western Blot
Classification Monoclonal
Clone OTI1D8
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Alpha-tubulin 6, bcm948, MGC10851, MGC14580, TUBA6, tubulin alpha-1C chain, Tubulin alpha-6 chain, tubulin, alpha 1c, tubulin, alpha 6
Gene Symbols TUBA1C
Host Species Mouse
Immunogen Full length recombinant protein of human TUBA6 (NP_116093) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84790
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.