Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                TUBA8 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
| Antigen | TUBA8 | 
|---|---|
| Clone | OTI3E10 | 
| Concentration | 0.63 mg/mL | 
| Classification | Monoclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        50-254-2609
                    
                    
                        
                    
                        
                    
                        
                    
                    
                        ![]()  
             
             | 
            
                
                    
                        OriGene
                         TA501009S  | 
                
            
            
            
            
            
                
                    30 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $231.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        50-167-2152
                    
                    
                        
                    
                        
                    
                    
                        ![]()  
             | 
            
                
                    
                        OriGene
                         TA501009  | 
                
            
            
            
            
            
                
                    100 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $516.29
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
TUBA8 Monoclonal antibody specifically detects TUBA8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TUBA8 | |
| 0.63 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| Primary | 
| OTI3E10 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| IgG2a | |
| -20° C, Avoid Freeze/Thaw Cycles | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title