Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUBB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110534 Supplier Invitrogen™ Supplier No. PA5110534
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMAATFIGNN TAIQEIFNRV SEHFSAMFKR KAFVHWYTSE GMDINEFGEA ENNIHDLVSE YQQFQDAKAV LEEDEEVTEE AEMEPEDKGH.

TUBB1 encodes a member of the beta tubulin protein family. Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. This protein is specifically expressed in platelets and megakaryocytes and may be involved in proplatelet production and platelet release. A mutations in this gene is associated with autosomal dominant macrothrombocytopenia. Two pseudogenes of this gene are found on chromosome Y.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TUBB1
Gene Accession No. A2AQ07
Gene Alias 2810484G07Rik; M(beta)1; tbb1; TUBB1; tubulin beta; tubulin beta 1 class VI; tubulin beta-1 chain; tubulin, beta 1 class VI
Gene Symbols TUBB1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 362-451 of human Tubulin beta-1 chain (NP_1104001).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 545486
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.