Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubby Antibody (SR2068), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32245850UL
Description
Tubby Monoclonal antibody specifically detects Tubby in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Tubby | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2068 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
rd5, tubby (mouse) homolog, tubby homolog (mouse), tubby homologue, tubby protein homolog | |
A synthesized peptide derived from human Tubby (Uniprot #: P50607) | |
50 μg | |
Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuroscience, Transcription Factors and Regulators | |
7275 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction