Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubby Antibody (SR2068), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
| Antigen | Tubby |
|---|---|
| Clone | SR2068 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
Description
Tubby Monoclonal antibody specifically detects Tubby in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Tubby | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| rd5, tubby (mouse) homolog, tubby homolog (mouse), tubby homologue, tubby protein homolog | |
| A synthesized peptide derived from human Tubby (Uniprot #: P50607) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2068 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuroscience, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7275 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title