Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119487
This item is not returnable.
View return policy
Description
TUBD1 Polyclonal Antibody for Western Blot, IHC (P)
Acts as a positive regulator of hedgehog signaling and regulates ciliary function. [UniProt].
Specifications
TUBD1 | |
Polyclonal | |
Unconjugated | |
TUBD1 | |
4930550G19Rik; Delta-tubulin; Tubd; TUBD1; tubulin delta 1; tubulin delta chain; tubulin, delta 1; tubulin, delta 1-like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51174 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UJT1 | |
TUBD1 | |
Recombinant protein Tubulin Delta. The antigen corresponds to amino acid range 190-453 of the target protein. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction