Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | TUBD1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119487
|
Invitrogen™
PA5119487 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121516
|
Invitrogen™
PA5121516 |
20 μL |
Each for $209.50
|
|
|||||
Description
TUBD1 Polyclonal Antibody for Western Blot, IHC (P)
Acts as a positive regulator of hedgehog signaling and regulates ciliary function. [UniProt].Specifications
TUBD1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
TUBD1 | |
4930550G19Rik; Delta-tubulin; Tubd; TUBD1; tubulin delta 1; tubulin delta chain; tubulin, delta 1; tubulin, delta 1-like | |
TUBD1 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9UJT1 | |
51174 | |
Recombinant protein Tubulin Delta. The antigen corresponds to amino acid range 190-453 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title