Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Tubes and Flat Caps PROMO
Click to view available options
:
Thermo Scientific™ Tubes and Caps
Description
Highlights
- Compatible with 0.2 mL thermal cycler blocks
- 8 tubes per strip
- 0.25 mL maximum tube capacity when capped
- Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications
Specifications
Certifications/Compliance | Certified DNA/RNase Free |
Color | Natural |
Volume (Metric) Working | 0.2 mL |
Volume (Metric) | 0.25 mL |
Quantity | 250 Strips |
For Use With (Equipment) | Thermal Cyclers |
Material | Polypropylene |
Reaction Speed | Standard |
Closure Type | Flat Cap |
No. of Wells | 96 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction